Silver nitrate 0,5% in aqueous solution, Surgipath®

Supplier: Leica Biosystems

Synonyms: Silver mononitrateNitric acid silver saltLunar caustic

Surgipath®
38016SS12C
75810-374EA 323.68 USD
75810-374
Silver nitrate 0,5% in aqueous solution, Surgipath®
Silver nitrate

Component of the Leica Modified GMS Silver Kit.


The silver component of the mixed Methenamine/Borax silver working solution.

Formula: AgNO₃
MW: 169.87 g/mol
MDL Number: MFCD00003414
CAS Number: 7761-88-8
https://us.vwr.com/stibo/web/std.lang.all/06/45/16640645.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR