Methenamine-Borax solution, Surgipath® for histology
Supplier: Leica Biosystems
Surgipath®
38016SS12D
75810-376EA 118.54 USD
75810-376
Methenamine-Borax solution, Surgipath® for histology
Methenamine-Borax
Component of the Leica Modified GMS Silver Stain Kit
  • Premixed
  • For silver impregnation
Premixed and then added with the silver nitrate component for silver impregnation.