Malachite green oxalate solution, Surgipath®
|
Danger
|
Synonyms:
Bis (N-{4-[[4-(dimethylamino)phenyl](phenyl)methylene]cyclohexa-2,5-dien-1-ylidene}-N-methylmethanaminium) trioxalate
, N-[4-[[4-(Dimethylamino)phenyl]phenylmethylene]-2,5-cyclohexadien-1-ylidene]-N-methyl-methanaminium, ethanedioate, ethanedioate(2:2:1)
, Basic Green 4
, N,N,N',N'-Tetramethyl-4,4'-diaminotriphenylcarbenium oxalate
Component of the Acid Fast Bacillus Special, Acid Fast Bacillus Special, Alcian Blue Special Stain Kits
- Background stain
- Pale color
Stains the background staining a pale green to blue/green.
Formula:
C₅₂H₅₄N₄O₁₂ MW: 927.02 g/mol |
MDL Number:
MFCD00011766 CAS Number: 2437-29-8 UN: 2790 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...