Hydrochloric acid solution, Surgipath®
Supplier: Leica Biosystems
Synonyms:
Muriatic acid
Surgipath®
38016SS7B
75806-656EA
104.48
USD
75806-656
Hydrochloric acid solution, Surgipath®
Hydrochloric acid
Component of the Perl's Prussion Blue Iron Stain Kit.
- Causes ionic iron reaction
- Creates blue stain
Combined with Potassium Ferracyanide solution, causes ionic iron to react and produce a blue pigment.
Formula:
HCl MW: 36.46 g/mol |
MDL Number:
MFCD00011324 CAS Number: 7647-01-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...