Trichrome special stain kit (Gomori), Surgipath® (Green collagen)
Supplier: Leica Biosystems
Synonyms:
Gomori Trichrome
Trichrome stains are used to stain and identify muscle fibers, collagen and nuclei. They can be used to contrast skeletal, cardiac or smooth muscle.
- For connective tissues
- Results in contrasting stain
Stains connective tissue with Gomori’s Blue to provide a brilliant contrasting stain.
The Gomori Trichrome is a simplification of the more elaborate stains and combines the plasma stain (chromotrope 2R) and connective tissue with Gomori’s Blue to provide a brilliant contrasting stain.
The Gomoris Blue and Green Kits contain 500 mL each of Bouins Solution, 1% Acetic Acid, and Gomoris Blue or Green Solutions. Also included are 250 mL each of Weigerts Hematoxylin, Solution A and B.