Carbol fuchsin solution, Surgipath®

Danger

Surgipath®
38016SS8A
75806-660EA 188.66 USD
75806-660
Carbol fuchsin solution, Surgipath®
Carbol fuchsin

Component of the Acid Fast Stain Kit.


  • Fast stain kit
  • Stains acid-fast bacilli


Stains the acid fast bacilli dark red.

Formula: C₂₁H₂₂ClN₃
MW: 351.88 g/mol
MDL Number: MFCD00013129
CAS Number: 4197-24-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR