Surgipath® Acid Fast Bacillus Stain Kit, Leica
Supplier: Leica
Surgipath®
38016SS8
75806-658KT 360.02 USD
75806-658
Surgipath® Acid Fast Bacillus Stain Kit, Leica
Stains Histology Stains
Acid Fast Bacillus Stain Kit stains are used to detect and identify acid fast bacilli in tissue.
  • Used to detect bacilli in tissue
  • For identifying infected tissue
A primary function of this stain is to identify tuberculosis in lung tissue, as well as other bacterial bacillus infected tissue. Bacilli are rod-shaped bacterial organisms.
Ordering information: This kit contains 500 mL each of Carbol-Fuchsin Solution and Malachite Green Solution.