Would you like to log in again?
Surgipath® Apex Adhesive Microscope Slides, Leica
Supplier: Leica Biosystems
Surgipath®
3800085 3800086 3800083
The Apex Adhesive Slides portfolio consists of microscope slides made of white glass and coated with a proprietary adhesive coating for a positive charge, to promote tissue adhesion and to allow the slide to be more hydrophilic.
  • Available in Green, Lavender, and Orange
  • Super adhesive
  • Hydrophilic
This hydrophilicity provides a better base for homogeneous covering by aqueous stains and reagents leading to consistent full slide staining.
All slides in the Apex portfolio are excellent for demanding procedures such as standard IHC, Special Stains and HIER techniques. All slides are 1 x 3 x .04”.
ORDER
Expand All / Collapse All
Description Color Supplier No. VWR Catalog Number Unit Price Quantity
Apex Adhesive Microscope Slides Lavender 3800085 75806-642 Case of 1,440 Retrieving Restricted
Apex Adhesive Microscope Slides Green 3800083 75806-640 Case of 1,440 Retrieving Restricted
Apex Adhesive Microscope Slides Orange 3800086 75806-644 Case of 1,440 Retrieving Restricted
restricted In order to process your orders without delay, we request that you provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:
  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name and address
* Please note if your account is within the State of California two of these pieces of identification will be required.
VWR will not lift restrictions for residential shipping addresses.
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Details
Select the article for which you are looking for Certificate
Indicate the Lot Number. This field is mandatory.
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty