Would you like to log in again?
Surgipath® SeleTech Hematoxylin and Eosin Staining System, Leica
Supplier: Leica Biosystems
Surgipath®
3801570 3802918 3803598 3801615
SelecTech is a hematoxylin and eosin staining system consisting of Hematoxylin 560, Blue Buffer 8, Define, Alcoholic Eosin Y 515 and Eosin Trichrome 515.
  • Standardized reagent
  • Passes vigorous analytical chemistry standards
SelecTech standardizes the reagents and stains used in the hematoxylin and eosin staining procedure. The reagents and stains that make up this new system must pass vigorous analytical chemistry standards and performance tests.
SelecTech Ready-To-Use Blue Buffer 8 is available in concentrates as well as ready-to-use formulas. Maintains a constant pH of 8 to minimize tissue loss and create desired coloration.
SelecTech Ready-To-Use Define aq is available in concentrates as well as ready-to-use formulas. Define is a refinement product designed to create crisp, nuclear detail and cosistency.
The new formulas eliminate the inconvenience of pouring from concentrate stock bottles and making dilutions for working solutions.
Log in to see your contract pricing and availability.
  Remember me on this device
ORDER
Expand All / Collapse All
Description Size Supplier No. VWR Catalog Number Unit Price Quantity
SeleTech Hematoxylin 500 mL 3801570 75810-352 Each Retrieving Restricted
SeleTech Alcoholic Eosin Y 515 500 mL 3801615 75810-354 Each Retrieving Restricted
SeleTech Blue Buffer, Ready to Use (RTU) 4 x 1 gal. 3802918 75810-358 Case of 4 Retrieving Restricted
SeleTech Aqua Define, Ready to Use (RTU) 4 x 1 gal. 3803598 75810-360 Case of 4 Retrieving Restricted
restricted In order to process your orders without delay, we request that you provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:
  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name and address
* Please note if your account is within the State of California two of these pieces of identification will be required.
VWR will not lift restrictions for residential shipping addresses.
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Details
Select the article for which you are looking for Certificate
Indicate the Lot Number. This field is mandatory.
Enter Certificate Details
Select Product:
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty