Surgipath® Dissection Knives, Leica
Surgipath®
3802148 3802144 3802142
75810-280BX 304.34 USD
75810-280 75810-278 75810-282
Surgipath® Dissection Knives, Leica
These Disposable Dissection Knives feature a high carbon stainless steel blade cast permanently to an aluminum handle.
  • Available in 6, 8 , and 12 inch blade sizes
  • Hollow ground blade
  • For clean, non-stick sectioning
Individual knives are hollow-ground and can be easily sterilized using routine sterilization techniques.