Surgipath® One-Piece Microbiopsy Cassettes, Leica
Supplier: Leica
Surgipath®
3802765 3802759 3802771 3802767 3802775 3802753 3802773 3802761 3802769 3802777 3802755 3802757 3802763 3802751
75809-360CS 354.72 USD
75809-360 75809-350 75809-362 75809-340 75809-352 75809-342 75809-354 75809-344 75809-356 75809-346 75809-358 75809-336 75809-348 75809-338
Surgipath® One-Piece Microbiopsy Cassettes, Leica
Embedding Cassettes Biopsy Processing Cassettes
One-Piece Microbiopsy Cassettes are designed to process even the smallest tissue biopsies including bone marrow biopsies.
  • Available in 14 colors
  • 0.3 mm Pore Size
  • 50° writing surface
Special vent design eases block formation and promotes air bubble release when embedding.