Benzopurpurine 4B, Powder
Supplier: MP BIOMEDICALS (FKA ICN BIOMED
|
Warning
|
Synonyms:
CI 23500
, Direct Red 2
, Direct Red 3
, Benzopurpurin B
0219506925
IC19506925EA
77.36
USD
IC19506925
Benzopurpurine 4B, Powder
Benzopurpurine 4B
Benzopurpurin B is used as a biological stain.
Visual transition interval: From pH 1.3 (violet) to pH 4.2 (red)
Physical Appearance: Dark red to brown powder
Solubility: Soluble in water (1mg/ml-clear dark red solution)
Usage Statement: Research Use Only (RUO). Ready for CE IVD labeling of clinical applications.
Formula:
C₃₄H₂₈N₆O₆S₂ MW: 724.72 g/mol Melting Pt: 554 °F (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00012410 CAS Number: 992-59-6 EINECS: 213-594-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...