Boc-D-CYS(ACM)-OH ≥99%

Supplier: INDOFINE CHEMICAL COMPANY MS

044881GM
100281-378EA 184.38 USD
100281-378
Boc-D-CYS(ACM)-OH ≥99%
Boc-D-CYS(ACM)-OH
Formula: C₁₁H₂₀N₂O₅S
MDL Number: MFCD00076917
CAS Number: 138775-00-5
ADR: ,
https://us.vwr.com/stibo/web/std.lang.all/66/87/21286687.jpg https://us.vwr.com/stibo/web/std.lang.all/66/86/21286686.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR