2,2'-Bis(Trifluoromethyl)-4,4'-diiodobiphenyl ≥97%

Supplier: INDOFINE CHEMICAL COMPANY MS

Synonyms: 4,4'-Diiodo-2,2'-bis(trifluoromethyl)-1,1'-biphenyl

21023250MG
100284-828EA 351.77 USD
100284-828
2,2'-Bis(Trifluoromethyl)-4,4'-diiodobiphenyl ≥97%
2,2'-Bis(Trifluoromethyl)-4,4'-diiodobiphenyl
Formula: C₁₄H₆F₆I₂
MW: 541.99 g/mol
MDL Number: MFCD00236673
CAS Number: 89803-70-3
ADR: ,
https://us.vwr.com/stibo/web/std.lang.all/69/22/21286922.jpg https://us.vwr.com/stibo/web/std.lang.all/69/21/21286921.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR