5,7-Bis(trifluoromethyl)-3-cyano-2-(cyanomethyl)pyrazolo[1,5-A]pyrimidine ≥97%

Supplier: INDOFINE CHEMICAL COMPANY MS

2624071GM
100275-218EA 1052.69 USD
100275-218
5,7-Bis(trifluoromethyl)-3-cyano-2-(cyanomethyl)pyrazolo[1,5-A]pyrimidine ≥97%
5,7-Bis(trifluoromethyl)-3-cyano-2-(cyanomethyl)pyrazolo[1,5-A]pyrimidine
Formula: C₁₁H₃F₆N₅
Melting Pt: 138…140 °C
MDL Number: MFCD00141799
CAS Number: 338786-45-1
ADR: ,
https://us.vwr.com/stibo/web/std.lang.all/62/29/21286229.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR