3,5-Bis(trifluoromethyl)benzophenone ≥97%
Supplier: INDOFINE CHEMICAL COMPANY MS
Synonyms:
1,3-Di(trifluormethyl)-5-(phenylmethanoyl)benzene
, (3,5-Bis(trifluoromethyl)phenyl)(phenyl)methanone
0930885GM
100274-578EA
124.23
USD
100274-578
3,5-Bis(trifluoromethyl)benzophenone ≥97%
3,5-Bis(trifluoromethyl)benzophenone
Formula:
C₁₅H₈F₆O MW: 318.22 g/mol |
MDL Number:
MFCD00015510 CAS Number: 21221-93-2 ADR: , |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...