3,3'-Bis(trifluoromethyl)benzhydrol ≥97%

Supplier: INDOFINE CHEMICAL COMPANY MS

210171GM
100284-814EA 118.9 USD
100284-814
3,3'-Bis(trifluoromethyl)benzhydrol ≥97%
3,3'-Bis(trifluoromethyl)benzhydrol
Formula: C₁₅H₁₀F₆O
MDL Number: MFCD00039228
ADR: ,
https://us.vwr.com/stibo/web/std.lang.all/69/20/21286920.jpg https://us.vwr.com/stibo/web/std.lang.all/69/19/21286919.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR