N,O-Bis(trifluoroacetyl)hydroxylamine ≥97%

Supplier: INDOFINE CHEMICAL COMPANY MS

091895GM
100282-960EA 420 USD
100282-960
N,O-Bis(trifluoroacetyl)hydroxylamine ≥97%
N,O-Bis(trifluoroacetyl)hydroxylamine
Formula: C₄HF₆NO₃
Melting Pt: 59…60 °C
MDL Number: MFCD00013565
CAS Number: 684-78-6
ADR: ,
https://us.vwr.com/stibo/web/std.lang.all/68/14/21286814.jpg https://us.vwr.com/stibo/web/std.lang.all/68/15/21286815.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR