Bzl-L-Met-OMe·HCI ≥99% (by HPLC), crystalline powder

Supplier: INDOFINE CHEMICAL COMPANY MS

044591GM
100274-224EA 124.23 USD
100274-224
Bzl-L-Met-OMe·HCI ≥99% (by HPLC), crystalline powder
Bzl-L-Met-OMe·HCI

  • White crystalline powder
  • Store at 0-8 °C

Formula: C₁₃H₁₉NO₂S·HCl
Melting Pt: 91…100 °C
MDL Number: MFCD02684382
CAS Number: 402929-60-6
ADR: ,
https://us.vwr.com/stibo/web/std.lang.all/61/74/21286174.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR