Bzl-D-Ala-OMe·HCI ≥99% (by HPLC), crystalline powder
Supplier: INDOFINE CHEMICAL COMPANY MS
044301GM
100271-566EA
243.23
USD
100271-566
Bzl-D-Ala-OMe·HCI ≥99% (by HPLC), crystalline powder
Bzl-D-Ala-OMe·HCI
- White crystalline powder
- Store at 0-8 °C
Formula:
C₁₁H₁₅NO₂·HCl Melting Pt: 176…185 °C |
MDL Number:
MFCD02684374 CAS Number: 95071-12-8 ADR: , |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...