Benzenesulfonyl fluoride ≥99%
Supplier: INDOFINE CHEMICAL COMPANY MS
260995GM
100285-670EA
176.54
USD
100285-670
Benzenesulfonyl fluoride ≥99%
Benzenesulphonyl fluoride
Formula:
C₆H₅FO₂S MW: 160.17 Boiling Pt: 203…204 °C |
MDL Number:
MFCD00007412 CAS Number: 368-43-4 ADR: , |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...