Behenyl arachidate ≥99%

Supplier: INDOFINE CHEMICAL COMPANY MS

452022100MG
100296-388EA 303.38 USD
100296-388
Behenyl arachidate ≥99%
Behenyl arachidate
MDL Number: MFCD00056281
CAS Number: 42232-87-1
ADR: ,
https://us.vwr.com/stibo/web/std.lang.all/74/38/21287438.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR