5,6,7-Trimethoxyflavone ≥98%, crystalline powder

Supplier: INDOFINE CHEMICAL COMPANY MS

Synonyms: 5,6,7-Trimethoxy-2-phenylchromen-4-oneBaicalein trimethyl ether

B25150MG
100265-668EA 64.08 USD
100265-668
5,6,7-Trimethoxyflavone ≥98%, crystalline powder
5,6,7-Trimethoxyflavone

  • Creamy-white crystalline powder
  • Store in a cool, dry place

Formula: C₁₈H₁₆O₅
MW: 312.32 g/mol
Melting Pt: 164…167 °C
MDL Number: MFCD00017457
CAS Number: 973-67-1
ADR: ,
https://us.vwr.com/stibo/web/std.lang.all/56/53/21285653.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR