Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Abbemat 350 and 550 Automatic Refractometers, Anton Paar

Supplier: Anton Paar
97787 97789
Abbemat digital refractometers allow fast and non-destructive refractive index measurements.

  • Easy operation
  • Color LCD screen and membrane keys which are resistant to spillage and dirt and can be operated even when wearing gloves
  • A reminder for test measurements or adjustments can be set at user-defined intervals
  • Automatic warning if the sample volume is not large enough for a valid measurement
  • No sample preparation, readings within seconds
  • A quick wipe cleans the prism after each measurement
  • Compliance and traceability
  • Full compliance with 21 CFR Part 11
  • Compliant with international standards such as ASTM, ICUMSA, OIML, AOAC, DIN/ISO, FDA, ISI, JIS and pharmacopoeias
  • History and current status of results via the result memory
  • Adjustment and check history, check intervals and validity checks for each measurement
  • Liquids, pastes, polymers and solids
  • Turbid, colored or opaque samples
  • Liquids containing air bubbles or solid particles
  • No influence from humidity, temperature or vibrations
  • A wide range of scales are stored in the Abbemat for converting refractive index into concentrations, etc.

They are factory calibrated with official standards from the Physikalisch-Technische Bundesanstalt (PTB, National Metrology Institute of Germany).

Refractive index results from Abbemat 350 are accurate to ±0.0001 nD. All Abbemat 350 refractometers provide full compliance with 21 CFR Part 11, with user levels, audit trail, and forgery-proof data export.

Refractive index results from Abbemat 550 provides an accuracy of ±0.00002 nD. All Abbemat 550 refractometers provide full compliance with 21 CFR Part 11, with user levels, audit trail, and forgery-proof data export.

Certifications: CE Compliant.




 

ORDER

Expand All / Collapse All
Description Model Accuracy Supplier No. VWR Catalog Number Unit Price Quantity
Automatic Refractometer Abbemat 550 ± 0.00002 nD 97787 75802-804 Each Retrieving

Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.

Product available on GSA Advantage®
Automatic Refractometer Abbemat 350 ± 0.0001 nD 97789 75801-648 Each Retrieving

Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.

Product available on GSA Advantage®
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Scales Sugar scales, syrup and honey scales, alcohol scales, urine scales, serum scales, more than 70 scales available
Temperature Sensor Built - in Pt 100 sensor
Temperature Control Automatic Peltier temperature control between 4°C and 85°C
Interfaces RS-232, CAN-bus, 3 USB ports, Ethernet, VGA
Light Source LED light source, average lifetime > 100,000 h
Electrical AC 100 to 240 VAC, 50/60Hz, 120 VA max.
Display TFT Touch screen 6.4”, 640 X 480 Pixels
Weight 6.5 kg (14.3 lbs)
Length 300 mm (11.8")
Width 145 mm (5.7")
Height 330 mm (13")

ACCESSORIES

Expand All / Collapse All

Accessories for Abbemat 350 and 550 Automatic Refractometers, Anton Paar
Description Details Supplier No. VWR Catalog Number Unit Price Quantity
Pharma Qualification Documentation Price for paperwork only; Labor and travel charges will apply and are only quoted through Anton Paar 101434 75801-536 Each Retrieving
Sample Presser For measurement of solid samples 110284 75801-534 Each Retrieving

Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.

Ultra Pure Water Refractive Index Calibration Standard: Ultra Pure Water, PTB National Metrology Institute, 20 ml Vol. 92322 75801-522 Each (25ml) Retrieving

Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.

Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

Q and A

Product Questions & Answers

Sort by: Most Helpful | Newest | Oldest

Items Per Page: 5  10  20 
2  3  4  Next

Can't find an answer to your question?  Ask us a question

Ask us a question:

Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question

Q&A Guidelines & Tips

If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.

When writing your question or answer, please follow these guidelines:

  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English

All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s

We reserve the right not to post your question if it contains any of the following

  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty