Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Abbemat 200 Automatic Refractometer, Anton Paar

Supplier: Anton Paar
109980
The refractometers of the Abbemat Economy Line provide all the features required for accurate refractive index measurements at an economic price.

  • Easy operation
  • Abbemat 200 refractometers are ready to operate on delivery. Just take them out of the box and start to measure
  • All settings can be easily accessed via intuitive navigation
  • The color LCD screen and membrane keys are resistant to spillage and dirt and can be operated even when wearing gloves
  • Automatic warning if the sample volume is not large enough for a valid measurement
  • A quick wipe cleans the prism after each measurement
  • Liquids, pastes, polymers and solids
  • Turbid, colored or opaque samples
  • Liquids containing air bubbles or solid particles
  • No influence from humidity, temperature or vibrations
  • A wide range of methods are stored in the Abbemat 200 for converting refractive index into concentrations, etc.

With its preprogrammed methods and optimized design, Abbemat 200 is a truly universal refractometer covering nearly all applications. Dedicated industry solutions are no longer required. The Abbemat 200 refractometers provide fast and accurate measurement of refractive index or concentration in all industry branches, from pharmaceuticals, flavors and chemicals to beverages and food. The Abbemat 200 refractometers allow fast and non-destructive refractive index measurements. They are factory-calibrated with official standards from the National Metrology Institute of Germany (PTB, Physikalisch-Technische Bundesanstalt).

Certifications: CE Compliant.

Ordering information: An instruction manual, extenral power supply, power cord, magnetic sample cover, drying cartridge, 5 Pasteur pipettes (3 ml) are supplied with every instrument.




 

ORDER

Expand All / Collapse All
Description Model Supplier No. VWR Catalog Number Unit Price Quantity
Automatic Refractometer Abbemat 200 109980 75801-652 Each Retrieving

Temperature sensitive This Item is temperature sensitive and has specific temperature and storage requirements for shipping/delivery. Shipping duration cannot exceed 2 days.

Product available on GSA Advantage®
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Scales Sugar scales, syrup and honey scales, alcohol scales, urine scales, serum scales, more than 70 scales available.
Accuracy ± 0.0001 nD
Temperature Sensor Built - in Pt 100 sensor
Temperature Control Automatic Peltier temperature control between 10°C and 60°C
Interfaces 3 USB, RS-232
Light Source LED light source, average lifetime 100,000 h.
Electrical AC 100 to 240 VAC, 50/60Hz, 120 VA max.
Display LCD 3.5”, 320 X 240 Pixels
Weight 6.5 kg (14.3 lbs)
Length 300 mm (11.8")
Width 145 mm (5.7")
Height 330 mm (13")

ACCESSORIES

Expand All / Collapse All

Accessories
Description Details Supplier No. VWR Catalog Number Unit Price Quantity
Pharma Qualification Documentation Price for paperwork only; Labor and travel charges will apply and are only quoted through Anton Paar 101434 75801-536 Each Retrieving
Sample Presser For measurement of solid samples 110284 75801-534 Each Retrieving

Temperature sensitive This Item is temperature sensitive and has specific temperature and storage requirements for shipping/delivery. Shipping duration cannot exceed 2 days.

Ultra Pure Water Refractive Index Calibration Standard: Ultra Pure Water, PTB National Metrology Institute, 20 ml Vol. 92322 75801-522 Each (25ml) Retrieving

Temperature sensitive This Item is temperature sensitive and has specific temperature and storage requirements for shipping/delivery. Shipping duration cannot exceed 2 days.

Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty