Would you like to log in again?
Accessories
Supplier: Anton Paar
143960 143961 143946 143949 143947 143948
ORDER
Expand All / Collapse All
Description Details Supplier No. VWR Catalog Number Unit Price Quantity
Cell with Luer Filling Ports Sample Tube 100 mm: Stainless steel cell with Luer filling ports, wireless Toolmaster, 100 mm Path Length, 5 mm I.D., 2.0 ml Vol. 143946 75801-542 Each Retrieving
Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.
Product available on GSA Advantage®
Cell with Luer Filling Ports Sample Tube 10 mm: Stainless steel cell with Luer filling ports, wireless Toolmaster, 10 mm Path Length, 5 mm I.D., 0.2 ml Vol. 143948 75801-674 Each Retrieving
Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.
Product available on GSA Advantage®
Cell with Luer Filling Ports Sample Tube 2.5 mm: Stainless steel cell with Luer filling ports, wireless Toolmaster, 2.5 mm Path Length, 5 mm I.D., 0.05 ml Vol. 143949 75801-676 Each Retrieving
Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.
Product available on GSA Advantage®
Cell with Luer Filling Ports Sample Tube 100 mm: Hastelloy C-276, Luer filling ports, wireless Toolmaster, 100 mm Path Length, 3 mm I.D., 0.7 ml Vol. 143947 75801-672 Each Retrieving
Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.
Product available on GSA Advantage®
Quartz Control Plate 100°Z/34°OD, wireless Toolmaster, 100°Z/34°OD Reference Values 143961 75801-544 Each Retrieving
Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.
Product available on GSA Advantage®
Quartz Control Plate 50°Z/17°OD, wireless Toolmaster, 50°Z/17°OD Reference Values 143960 75801-678 Each Retrieving
Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.
Product available on GSA Advantage®
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Details
Select the article for which you are looking for Certificate
Indicate the Lot Number. This field is mandatory.
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty