H-Asn-OtBu ≥98%, powder

Supplier: INDOFINE CHEMICAL COMPANY MS

043251GM
100281-164EA 190.92 USD
100281-164
H-Asn-OtBu ≥98%, powder
H-Asn-OtBu

  • White to off white powder
  • Store at 0-8 °C

Formula: C₈H₁₆N₂O₃
MDL Number: MFCD00190755
CAS Number: 25456-86-4
ADR: ,
https://us.vwr.com/stibo/web/std.lang.all/66/64/21286664.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR