Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3-(4,4-difluoro-5,7-dimethyl-4-bora-3a,4a-diaza-s-indacene-3-yl)propionic acid NHS ester

Supplier: MARKER GENE TECHNOLOGIES MS

M2482
BODIPY®FL NHS Ester equivalent.

  • CAS: [146616-66-2]

Boron-dipyrromethene dyes (trade name BODIPY®) are composed of a dipyrrole structure containing a complexed boron difluoride. These fluorophores are characterized by a low molecular weight and small stokes shift, but with high quantum yields, sharp emission spectra and low photobleaching attributes that make them useful in a variety of biological analysis techniques. Their fluorescence is also predominantly environment-insensitive in a variety of different solvents or in different polarity situations in cells or tissues (e.g. in the lipid bilayer, cytosol, etc.). This dye has fluorescence excitation and emission properties (EX 503 nm; EM 512 nm) that are very similar to fluorescein and as such can be used with many current instrumentation systems.


Formula: C₁₈H₁₈BF₂N₃O₄
MW: 389,166 g/mol
CAS Number: 146616-66-2

Log in to see your contract pricing and availability.
  Remember me on this device
 

ORDER

Expand All / Collapse All
Size Supplier No. VWR Catalog Number Unit Price Quantity
5 mg M2482 102971-834 Each (5mg) Retrieving Restricted

Temperature sensitive This Item is temperature sensitive and has specific temperature and storage requirements for shipping/delivery. Requires a maximum 2 day delivery.

restricted In order to process your orders without delay, we request that you provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name and address

* Please note if your account is within the State of California two of these pieces of identification will be required.
VWR will not lift restrictions for residential shipping addresses.

Expand All / Collapse All
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Select Product:

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty