Tris(carbobenzoxy)-L-arginine ≥99%

Supplier: INDOFINE CHEMICAL COMPANY MS

Synonyms: Nα,Nδ,Nomega-Tris(benzyloxycarbonyl)-L-arginineZ-Arg(Z)2-OHTri-Cbz-L-arginine

0425405GM
100292-706EA 253.69 USD
100292-706
Tris(carbobenzoxy)-L-arginine ≥99%
Tris(carbobenzoxy)-L-arginine
Formula: C₃₀H₃₂N₄O₈
MW: 576.61 g/mol
Melting Pt: 137…140 °C
MDL Number: MFCD00038693
CAS Number: 14611-34-8
ADR: ,

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR