IBSMM0261
M0261
101176-234EA 62.31 USD
101176-234
IBSMM0261
β-Glucuronidase inhibitor similar to PETG.
This hydrophobic thio-glycoside can be used as a competitive inhibitor of β-glucuronidase activity.