Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Fluorescein-5-thiosemicarbazide HCl salt

Supplier: MARKER GENE TECHNOLOGIES MS
Synonyms: 5-FTSC HCl salt

M1036
Cell impermeant fluorescent probe useful for determining protein and peptide topology on the cell surface.

  • Cell impermeant probe

Can be reversibly coupled to aldehydes and ketones to form a Schiff base - which can be further reduced to a generate stable amine derivatives by treatment with reducing agents such as sodium borohydride (NaBH4) or sodium cyanoborohydride ((NaCNH3). Carboxylic acids of proteins and other water-soluble biopolymers can also be coupled to this fluorescein derivative in aqueous solution using water-soluble carbodiimides such as EDAC.


Formula: C₂₁H₁₅N₃O₅S·HCl
MW: 457,89 g/mol
CAS Number: 76863-28-0

 

ORDER

Expand All / Collapse All
Size Supplier No. VWR Catalog Number Unit Price Quantity
50 mg M1036 101447-508 Each (50mg) Retrieving Restricted
restricted In order to process your orders without delay, we request that you provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name and address

* Please note if your account is within the State of California two of these pieces of identification will be required.
VWR will not lift restrictions for residential shipping addresses.

Expand All / Collapse All
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty
 

A new look and a renewed commitment

Built on the features & resources you know, optimized with innovation from Avantor®. Our new brand symbolizes agility and our ability to adapt to customer needs.

Learn More Start Shopping