Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

N-Succinimidyl 6-Biotinamidohexanoate

Supplier: MARKER GENE TECHNOLOGIES MS
Synonyms: 6-Biotinamidohexanoic Acid N-Succinimidyl EsterN-Biotinyl-6-aminohexanoic Acid Succinimidyl EsterN-[5-(N-Succinimidyloxycarbonyl)pentyl]biotinamide

M0783
This reactive biotin derivative contains an aminohexanoyl spacer group (termed "X") between biotin and the binding site for proteins, nucleic acids, or other biological molecules which helps to reduce potential quenching effects of fluorophores attached to the analyte molecule, since biotin binds to "deep" binding sites in avidin or streptavidin.

  • Protein label
  • Spacer "X" reduces quenching effects

The extra linking arm of this biotin derivative makes it more accessible for binding, and improves a variety of assays employing biotin-avidin or biotin-streptavidin systems. This amine reactive (succinimidyl ester, SE) derivative can be readily coupled to primary or secondary amines of analyte compounds or proteins under mild conditions. Biotin is a commonly used hapten that can be detected by using its strong affinity for the proteins avidin (from egg white) or streptavidin. Many avidin or streptavidin conjugates are available that have been fluorescently labeled or are conjugated to an antibody or other detection reagent. This biotin labeling compound has been used in many such detection schemes for peptides, proteins, organelle components, monoclonal antibody analyses, oligonucleotide and microbead labelings


Formula: C₂₀H₃₀N₄O₆S
MW: 454.54 g/mol
MDL Number: MFCD00065502
CAS Number: 72040-63-2

Log in to see your contract pricing and availability.
  Remember me on this device
 

ORDER

Expand All / Collapse All
Size Supplier No. VWR Catalog Number Unit Price Quantity
50 mg M0783 101176-324 Each (50mg) Retrieving Restricted
restricted In order to process your orders without delay, we request that you provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name and address

* Please note if your account is within the State of California two of these pieces of identification will be required.
VWR will not lift restrictions for residential shipping addresses.

Expand All / Collapse All
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty