3-(Trifluoromethyl)anthranilic acid ≥98%

Supplier: INDOFINE CHEMICAL COMPANY MS

2629975GM
100287-148EA 121.62 USD
100287-148
3-(Trifluoromethyl)anthranilic acid ≥98%
3-(Trifluoromethyl)anthranilic acid
Formula: C₈H₆F₃NO₂
MW: 205.14 g/mol
Melting Pt: 157…160 °C
MDL Number: MFCD00274210
CAS Number: 313-12-2
ADR: ,
https://us.vwr.com/stibo/web/std.lang.all/69/95/21286995.jpg https://us.vwr.com/stibo/web/std.lang.all/69/94/21286994.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR