4-(3-(Trifluoromethyl)-1H-pyrazol-1-yl)aniline ≥97%
Supplier: INDOFINE CHEMICAL COMPANY MS
Synonyms:
1-(4-Aminophenyl)-3-(trifluoromethyl)pyrazole
260661GM
100272-148EA
376.62
USD
100272-148
4-(3-(Trifluoromethyl)-1H-pyrazol-1-yl)aniline ≥97%
4-(3-(Trifluoromethyl)-1H-pyrazol-1-yl)aniline
MW:
227.19 g/mol |
MDL Number:
MFCD00115025 CAS Number: 231953-33-6 ADR: , |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...