1-Aminocyclopropanecarboxylic acid ≥99% (by titrimetric analysis), Powder
Supplier: INDOFINE CHEMICAL COMPANY MS
Synonyms:
1-Amino-1-cyclopropanecarboxylic acid
042645GM
100277-022EA
122.92
USD
100277-022
1-Aminocyclopropanecarboxylic acid ≥99% (by titrimetric analysis), Powder
1-Aminocyclopropanecarboxylic acid
- White powder
- Store at 0-8 °C
Formula:
C₄H₇NO₂ MW: 101.11 g/mol |
MDL Number:
MFCD00009944 CAS Number: 22059-21-8 ADR: , |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...