4-Amino-2-cyano-3'-(trifluoromethyl)diphenyl thioether ≥95%

Supplier: INDOFINE CHEMICAL COMPANY MS

2622221GM
100286-506EA 346.54 USD
100286-506
4-Amino-2-cyano-3'-(trifluoromethyl)diphenyl thioether ≥95%
4-Amino-2-cyano-3'-(trifluoromethyl)diphenyl thioether
Formula: C₁₄H₉F₃N₂S
Melting Pt: 78…80 °C
MDL Number: MFCD00793554
CAS Number: 320421-51-0
ADR: ,
https://us.vwr.com/stibo/web/std.lang.all/69/78/21286978.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR