Z-ABU-OSu ≥98%

Supplier: INDOFINE CHEMICAL COMPANY MS

042551GM
100280-844EA 196.15 USD
100280-844
Z-ABU-OSu ≥98%
Z-ABU-OSu
Formula: C₁₆H₁₈N₂O₆
MDL Number: MFCD00191058
CAS Number: 71447-81-9
ADR: ,
https://us.vwr.com/stibo/web/std.lang.all/66/48/21286648.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR