H-2-Abu-OtBu·HCl ≥99% (by HPLC), powder

Supplier: INDOFINE CHEMICAL COMPANY MS

Synonyms: tert-Butyl (S)-2-aminobutyrate hydrochlorideH-Abu-OtBu·HCl

042521GM
100271-454EA 102 USD
100271-454
H-2-Abu-OtBu·HCl ≥99% (by HPLC), powder
H-2-Abu-OtBu·HCl

  • White powder
  • Store at 0-8 °C

Formula: C₈H₁₈ClNO₂
MW: 195.687 g/mol
Melting Pt: 240…245 °C
MDL Number: MFCD00237697
CAS Number: 53956-05-1
ADR: ,
https://us.vwr.com/stibo/web/std.lang.all/60/27/21286027.jpg https://us.vwr.com/stibo/web/std.lang.all/60/28/21286028.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR