(S)-3-(Allylsulfinyl)-L-alanine ≥98% (by HPLC), powder

Supplier: INDOFINE CHEMICAL COMPANY MS

027253S10MG
100266-540EA 328.23 USD
100266-540
(S)-3-(Allylsulfinyl)-L-alanine ≥98% (by HPLC), powder
(S)-3-(Allylsulfinyl)-L-alanine

  • White powder
  • To be kept in desiccator

Formula: C₆H₁₁NO₃S
Melting Pt: 152…153 °C
MDL Number: MFCD01311056
CAS Number: 556-27-4
ADR: ,
https://us.vwr.com/stibo/web/std.lang.all/57/21/21285721.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR