β-Alanine ≥98%, crystalline powder

Supplier: INDOFINE CHEMICAL COMPANY MS

Synonyms: 3-Aminopropanoic acidH-β-Ala-OHH-Bala-OH3-Aminopropionic acid

025502250GM
100263-070EA 115.08 USD
100263-070
β-Alanine ≥98%, crystalline powder
β-Alanine

  • White crystalline powder
  • Store in a cool, dry place

Formula: C₃H₇NO₂
MW: 89.09 g/mol
Melting Pt: 205 °C (dec.)
MDL Number: MFCD00008200
CAS Number: 107-95-9
ADR: ,
https://us.vwr.com/stibo/web/std.lang.all/55/36/21285536.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR