Ajmalicine ≥95% (by HPLC)

Supplier: INDOFINE CHEMICAL COMPANY MS

02169210MG
100264-728EA 221 USD
100264-728
Ajmalicine ≥95% (by HPLC)
Ajmalicine
Formula: C₂₁H₂₄N₂O₃
MDL Number: MFCD00042748
CAS Number: 483-04-5
ADR: ,

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR