Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

N-Acetyl-D-tyrosine ≥99%

Supplier: INDOFINE CHEMICAL COMPANY MS
Synonyms: Ac-D-Tyr-OH

041055GM
100278-656EA 555.77 USD InStock
100278-656
N-Acetyl-D-tyrosine ≥99%
N-Acetyl-D-tyrosine


Formula: C₁₁H₁₃NO₄
MW: 223.228 g/mol
MDL Number: MFCD00063037
CAS Number: 19764-32-0
UN: 12352100
ADR: ,
https://us.vwr.com/stibo/web/std.lang.all/65/09/21286509.jpg
Order Now

 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty