Ac-Phe-OMe ≥99%

Supplier: Avantor

Synonyms: Methyl N-acetyl-L-phenylalaninate

040925GM
100272-572EA 94 USD
100272-572
Ac-Phe-OMe ≥99%
Ac-Phe-OMe
Formula: C₁₂H₁₅NO₃
MW: 221.256 g/mol
MDL Number: MFCD00066060
CAS Number: 3618-96-0
ADR: ,
https://us.vwr.com/stibo/web/std.lang.all/65/00/21286500.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR