N-Acetyl-neuraminic acid methyl ester ≥98%

Supplier: INDOFINE CHEMICAL COMPANY MS

561151100MG
100291-404EA 604.15 USD
100291-404
N-Acetyl-neuraminic acid methyl ester ≥98%
N-Acetyl-neuraminic acid methyl ester
MDL Number: MFCD00214229
CAS Number: 50998-13-5
ADR: ,
https://us.vwr.com/stibo/web/std.lang.all/71/34/21287134.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR