Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Dichloromethane ≥99.8% CHROMASOLV™ for pesticide residue analysis

Supplier: Honeywell Research Chemicals
Synonyms: Methane dichlorideMethylene chloride

Stabilization: Stabilised with amylene

34488-4X2.5L 34488-4X4L 34488-6X1L
Dichloromethane (DCM, Methylene chloride) is an organic compound with formula CH2Cl2. This colorless, volatile liquid has a moderately sweet odor and is widely used as a solvent. Although it is not miscible with water, it is miscible with many organic solvents. Suitable for Pesticide Resisdue Analysis (PRA).


Formula: CH₂Cl₂
MW: 84.93 g/mol
Boiling Pt: 39,8-40 °C (lit.)
Melting Pt: −97 °C (lit,)
Density: 1,325 g/mL at 25 °C (lit.)
Storage Temperature: Ambient
MDL Number: MFCD00000881
CAS Number: 75-09-2
EINECS: 200-838-9
UN: 1593
ADR: 6.1,III
https://us.vwr.com/stibo/web/std.lang.all/14/30/20961430.jpg


 

ORDER

Expand All / Collapse All
Size Packaging Supplier No. VWR Catalog Number Unit Price Quantity
1 L Glass Bottle 34488-6X1L BJ34488-1L Case of 6 (1 l) Retrieving Restricted
Each(1l) Retrieving Restricted
4 L Glass Bottle 34488-4X4L BJ34488-4L Case of 4 (4 l) Retrieving Restricted
Each(4l) Retrieving Restricted
2.5 L Glass Bottle 34488-4X2.5L BJ34488-2.5L Case of 4 (2,500 ,ml) Retrieving Restricted
Each(2,500,ml) Retrieving Restricted

Temperature sensitive This Item is temperature sensitive and has specific temperature and storage requirements for shipping/delivery. Shipping duration cannot exceed 2 days.

restricted In order to process your orders without delay, we request that you provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name and address

* Please note if your account is within the State of California two of these pieces of identification will be required.
VWR will not lift restrictions for residential shipping addresses.

Expand All / Collapse All
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty