Ethyl-N-acetylglycinate(Ac-Gly-OEt) ≥98% (by HPLC), crystalline powder

Supplier: INDOFINE CHEMICAL COMPANY MS

Synonyms: Ac-Gly-OEt

04047100GM
100294-430EA 258.92 USD
100294-430
Ethyl-N-acetylglycinate(Ac-Gly-OEt) ≥98% (by HPLC), crystalline powder
Ethyl N-acetylglycinate (Ac-Gly-OEt)

  • White or off-white crystalline powder
  • Store at 0-8 °C

Formula: C₆H₁₁NO₃
MW: 145.1 g/mol
Melting Pt: 42…48 °C
MDL Number: MFCD00009173
CAS Number: 1906-82-7
ADR: ,
https://us.vwr.com/stibo/web/std.lang.all/73/21/21287321.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR