Acetyl-(D3)-L-carnitine.HCl

Supplier: INDOFINE CHEMICAL COMPANY MS

71173210MG
100291-768EA 895.77 USD
100291-768
Acetyl-(D3)-L-carnitine.HCl
Acetyl-(D3)-L-carnitine.HCl
MDL Number: MFCD06656338
CAS Number: 362049-62-5
ADR: ,
https://us.vwr.com/stibo/web/std.lang.all/71/77/21287177.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR