Would you like to log in again?
Global Web is currently experiencing issues, technical teams are working to correct the problem.
Modular WorkSafe Rubber Flooring, Wearwell®
Supplier: WEARWELL
472.12X3X3GPBK 472.12X3X3GRBK
Create a safe, ergonomically designed mat that works in wet, dry, or even greasy environments.
  • Lightweight and easy to clean
  • Hidden connections require no additional attachments
  • Safety rounded edges
Designed for comfort and ease of use, Modular WorkSafe Rubber Flooring is popular in high maintenance areas. Choose between general-purpose GR natural rubber for dry areas and exposure to water, or NBR for areas with petrochemicals.
Certifications: Meets ASTM F1677. Dry: COF = >0.84/Wet: COF = 0.78/Oily: COF = 0.34. Meets ASTM D575. Deflection @35 psi, mm = 4.10.
Log in to see your contract pricing and availability.
  Remember me on this device
Expand All / Collapse All
Description Length Thickness Width Color Supplier No. VWR Catalog Number Unit Price Quantity
Modular WorkSafe Light Grease Proof Mat 3' 0.5" 3' Black 472.12X3X3GPBK 89020-970 Each Retrieving
Modular WorkSafe Light Grease Resistant Mat 3' 0.5" 3' Black 472.12X3X3GRBK 89020-972 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Details
Select the article for which you are looking for Certificate
Indicate the Lot Number. This field is mandatory.
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty