Riboflavine (vitamin B2) 99.0-101.0%, Calbiochem®
Supplier: MilliporeSigma
Synonyms:
Lactoflavin
, 7,8-Dimethyl-10-((2S,3S,4R)-2,3,4,5-tetrahydroxypentyl)benzo[g]pteridine-2,4(3H,10H)-dione
, Vitamin B2
, Vitamine G
, (-)-Riboflavin
555682-10GM
80109-776EA
67.4
USD
80109-776
Riboflavine (vitamin B2) 99.0-101.0%, Calbiochem®
Riboflavine (Vitamin B2)
A water soluble vitamin involved in the synthesis of flavin mononucleotide (FMN) and flavin adenine dinucleotide (FAD). Provides protection against oxidative damage.
Formula:
C₁₇H₂₀N₄O₆ MW: 376.37 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00005022 CAS Number: 83-88-5 |
Specification Test Results
Synonyms | Vitamin B₂ |
Form | Yellow-Orange Solid |
Hill Formula | C17H20N4O6 |
Chemical Formula | C17H20N4O6 |
Optical Rotation | -135.0° to -155.0° (c = 0.5%, 50 mM NaOH, dry basis) |
Purity | 99.0 - 101.0% by UV |
Contaminants | Conforms (Lumiflavine) |
Toxicity | Standard Handling |
Storage | 15 to 30 °C |
Protect from Light | Protect from Light |
Do not Freeze | Ok to Freeze |
Special Instructions | Following reconstitution, aliquot and freeze (-20 °C). Stock solutions are stable for up to 3 months at -20 °C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...