6-Acetyl-2-Benzoxazolinone ≥99%, powder
Supplier: INDOFINE CHEMICAL COMPANY MS
Synonyms:
6-Acetylbenzo[d]oxazol-2(3H)-one
250021GM
100267-850EA
52.31
USD
100267-850
6-Acetyl-2-Benzoxazolinone ≥99%, powder
6-Acetyl-2-Benzoxazolinone
- Light browish cream color powder
- Store in a cool, dry place
Formula:
C₉H₇NO₃ MW: 177.16 g/mol Melting Pt: 233…235 °C |
MDL Number:
MFCD01664312 CAS Number: 54903-09-2 ADR: , |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...